Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 2 | 290 | ||
GenBank mRNA | AB009685.1 | - | 1 | 86 | ||
AF060923.1 | - | 2 | 164 | |||
AF078863.1 | - | 2 | 215 | |||
AF131820.1 | - | 2 | 217 | |||
AF161400.1 | - | 2 | 199 | |||
AK098770.1 | - | 2 | 163 | |||
AK130094.1 | - | 2 | 193 | |||
AK130097.1 | - | 2 | 158 | |||
AK225318.1 | - | 2 | 167 | |||
AK292864.1 | - | 2 | 158 | |||
AK297960.1 | - | 2 | 157 | |||
AK298332.1 | - | 2 | 163 | |||
AK299937.1 | - | 196 | ||||
AL136713.1 | - | 2 | 200 | |||
AY358824.1 | - | 2 | 165 | |||
BC010354.1 | - | 2 | 161 | 4093037 | ||
BC022872.1 | - | 2 | 172 | |||
CR533519.1 | - | 1 | 86 |