Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 6 | 775 | ||
GenBank mRNA | AB009685.1 | - | 5 | 571 | ||
AF060923.1 | - | 6 | 649 | |||
AF078863.1 | - | 6 | 700 | |||
AF131820.1 | - | 6 | 702 | |||
AK098770.1 | - | 7 | 588 | |||
AK130094.1 | - | 6 | 678 | |||
AK130097.1 | - | 6 | 643 | |||
AK225318.1 | - | 6 | 652 | |||
AK292864.1 | - | 6 | 643 | |||
AK297960.1 | - | 6 | 568 | |||
AK298332.1 | - | 6 | 657 | |||
AK299937.1 | - | 5 | 681 | |||
AY358824.1 | - | 6 | 650 | |||
BC010354.1 | - | 6 | 646 | 4093037 | ||
BC022872.1 | - | 6 | 657 |