Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198558.1 | NP_001185487.1 | 1, 2 | 506 | ||
NM_014441.2 | NP_055256.1 | 1, 2 | 506 | |||
XM_006723146.3 | XP_006723209.1 | 1, 2 | 478 | |||
XM_011526730.2 | XP_011525032.1 | 1, 2 | 478 | |||
XM_011526732.2 | XP_011525034.1 | 1, 2 | 478 | |||
XM_017026595.1 | XP_016882084.1 | 1, 2 | 478 | |||
XM_017026596.1 | XP_016882085.1 | 1, 2 | 478 | |||
GenBank mRNA | AB026265.1 | - | 1, 2 | 457 | ||
AF227924.1 | - | 1, 2 | 478 | |||
AF247180.1 | - | 1, 2 | 422 | |||
AK297043.1 | - | 1, 2 | 478 | |||
AK315272.1 | - | 1, 2 | 449 | |||
AY358913.1 | - | 1, 2 | 453 | |||
BC035365.1 | - | 1, 2 | 460 | 5176457 |