Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014469.4 | NP_055284.3 | 1 | 403 | ||
GenBank mRNA | AF069682.2 | - | 1 | 309 | ||
AK292271.1 | - | 1 | 310 | |||
BC029295.1 | - | 1 | 228 | |||
BC057796.1 | - | 1 | 234 | 5261317 |