Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 24 | 5088 | ||
NM_001252612.1 | NP_001239541.1 | 24 | 5414 | |||
NM_001252613.1 | NP_001239542.1 | 24 | 5098 | |||
NM_014497.4 | NP_055312.2 | 24 | 5098 | |||
XM_005264263.1 | XP_005264320.1 | 23 | 3562 | |||
XM_006711989.1 | XP_006712052.1 | 23 | 3574 | |||
XM_011532768.2 | XP_011531070.2 | 23 | 5058 | |||
XM_017003809.1 | XP_016859298.1 | 24 | 5189 | |||
XM_017003810.1 | XP_016859299.1 | 23 | 4973 | |||
GenBank mRNA | AF273049.1 | - | 14 | 2531 | ||
AF534078.1 | - | 22 | 3553 | |||
AK093578.1 | - | 3 | 669 | |||
AK128706.1 | - | 5 | 3055 | |||
BC083513.1 | - | 24 | 5057 | 5540321 | ||
BC143728.1 | - | 24 | 5051 | 9052242 | ||
BX537425.1 | - | 24 | 5062 | |||
CR749322.1 | - | 24 | 5411 | |||
D83032.1 | - | 24 | 5090 |