Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005618.3 | NP_005609.3 | 10 | 2555 | ||
XM_005266934.3 | XP_005266991.1 | 10 | 2424 | |||
GenBank mRNA | AF196571.1 | - | 10 | 2403 | ||
AK314234.1 | - | 10 | 2427 | |||
AY358892.1 | - | 10 | 2122 |