Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014035.2 | NP_054754.1 | 3 | 382 | ||
XM_005271972.4 | XP_005272029.1 | 3 | 271 | |||
XM_006714592.3 | XP_006714655.1 | 3 | 337 | |||
XM_011543349.2 | XP_011541651.1 | 4 | 402 | |||
XM_011543350.2 | XP_011541652.1 | 4 | 401 | |||
XM_011543352.1 | XP_011541654.1 | 4 | 402 | |||
XM_017009394.1 | XP_016864883.1 | 3 | 207 | |||
XM_017009395.1 | XP_016864884.1 | 4 | 402 | |||
XM_017009396.1 | XP_016864885.1 | 4 | 402 | |||
GenBank mRNA | AF139461.1 | - | 3 | 271 | ||
AY044655.1 | - | 3 | 213 | |||
AY358098.1 | - | 3 | 231 | |||
BC010886.1 | - | 3 | 342 | 3538426 | ||
BC069012.1 | - | 3 | 329 | 4585781 | ||
BQ278046.1 | - | 3 | 354 |