Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 5 | 414 | ||
NM_001258440.1 | NP_001245369.1 | 5 | 354 | |||
NM_001258441.1 | NP_001245370.1 | 5 | 354 | |||
NM_001258442.1 | NP_001245371.1 | 5 | 354 | |||
NM_013314.3 | NP_037446.1 | 5 | 414 | |||
NR_047680.1 | 5 | 354 | ||||
NR_047681.1 | 5 | 359 | ||||
NR_047682.1 | 5 | 354 | ||||
NR_047683.1 | 5 | 354 | ||||
XM_011539728.2 | XP_011538030.1 | 5 | 291 | |||
XM_017016159.1 | XP_016871648.1 | 5 | 292 | |||
GenBank mRNA | AF068180.1 | - | 5 | 389 | ||
AF068181.1 | - | 5 | 389 | |||
AK027779.1 | - | 5 | 349 | |||
AK225546.1 | - | 5 | 397 | |||
AM180327.1 | - | 5 | 236 | |||
AM180328.1 | - | 5 | 236 | |||
AM180329.1 | - | 5 | 236 | |||
AM180330.1 | - | 5 | 236 | |||
AM180331.1 | - | 5 | 236 | |||
AM180332.1 | - | 5 | 236 | |||
AM180333.1 | - | 5 | 236 | |||
AM180334.1 | - | 5 | 236 | |||
AM180336.1 | - | 5 | 236 | |||
AM180337.1 | - | 5 | 236 | |||
AM180338.1 | - | 5 | 241 | |||
AM180339.1 | - | 5 | 236 | |||
AM180340.1 | - | 5 | 236 | |||
AM180341.1 | - | 4 | 186 | |||
AM180342.1 | - | 4 | 186 | |||
AM180343.1 | - | 4 | 186 | |||
AM180344.1 | - | 3 | 145 | |||
AM180345.1 | - | 4 | 186 | |||
AM180346.1 | - | 3 | 184 | |||
BC018906.2 | - | 5 | 328 | 4053894 | ||
BC110298.1 | - | 5 | 346 | |||
BM820310.1 | - | 5 | 407 | |||
BU597768.1 | - | 5 | 359 | |||
DA045252.1 | - | 5 | 414 |