Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAZ2B SIRNA Assays ›
Gene Symbol:
BAZ2B
Gene Name:
bromodomain adjacent to zinc finger domain 2B
Gene Aliases:
WALp4
Chromosome Location:
Chr.2: 159315312 - 159712439 on Build GRCh38
UniGene ID:
Hs.470369
Species:
Human
siRNA ID
HSS121183
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAZ2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013450.3
NP_038478.2
13
2814
XM_005246488.2
XP_005246545.2
13
2657
XM_005246489.4
XP_005246546.2
12
3760
XM_005246492.4
XP_005246549.2
12
3755
XM_006712461.3
XP_006712524.2
16
3231
XM_011511038.2
XP_011509340.1
13
2761
XM_011511039.1
XP_011509341.1
11
2411
XM_011511042.1
XP_011509344.1
11
2411
XM_011511045.1
XP_011509347.1
10
2216
XM_011511047.1
XP_011509349.1
12
2563
XM_011511048.2
XP_011509350.1
12
2628
XM_011511050.1
XP_011509352.1
10
2111
XM_011511053.2
XP_011509355.1
11
2411
XM_011511054.2
XP_011509356.1
11
2411
XM_017003918.1
XP_016859407.1
16
3221
XM_017003919.1
XP_016859408.1
16
3215
XM_017003920.1
XP_016859409.1
18
3432
XM_017003921.1
XP_016859410.1
13
2755
XM_017003922.1
XP_016859411.1
13
2651
XM_017003923.1
XP_016859412.1
11
2411
XM_017003926.1
XP_016859415.1
12
2523
XM_017003927.1
XP_016859416.1
12
2563
XM_017003928.1
XP_016859417.1
12
2467
XM_017003930.1
XP_016859419.1
12
2363
XM_017003931.1
XP_016859420.1
12
2461
XM_017003933.1
XP_016859422.1
11
2334
XM_017003934.1
XP_016859423.1
11
2229
GenBank mRNA
AB032255.1
-
8
2187
AK127551.1
-
12
2224
AK307683.1
-
11
2051
AL834381.1
-
13
2709
BC012576.1
-
9
1827
BX641037.1
-
16
5623
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0