Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAZ2B SIRNA Assays ›
Gene Symbol:
BAZ2B
Gene Name:
bromodomain adjacent to zinc finger domain 2B
Gene Aliases:
WALp4
Chromosome Location:
Chr.2: 159315312 - 159712439 on Build GRCh38
UniGene ID:
Hs.470369
Species:
Human
siRNA ID
HSS121185
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAZ2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289975.1
NP_001276904.1
29, 30
5584
NM_013450.3
NP_038478.2
30, 31
5692
XM_005246488.2
XP_005246545.2
31, 32
5568
XM_005246489.4
XP_005246546.2
30, 31
6671
XM_005246492.4
XP_005246549.2
30, 31
6666
XM_006712461.3
XP_006712524.2
34, 35
6142
XM_011511038.2
XP_011509340.1
31, 32
5666
XM_011511039.1
XP_011509341.1
28, 29
5289
XM_011511042.1
XP_011509344.1
29, 30
5322
XM_011511043.1
XP_011509345.1
27, 28
5187
XM_011511044.1
XP_011509346.1
28, 29
5214
XM_011511045.1
XP_011509347.1
28, 29
5127
XM_011511047.1
XP_011509349.1
30, 31
5474
XM_011511048.2
XP_011509350.1
30, 31
5539
XM_011511050.1
XP_011509352.1
28, 29
5022
XM_011511051.1
XP_011509353.1
28, 29
5208
XM_011511052.1
XP_011509354.1
27, 28
4926
XM_011511053.2
XP_011509355.1
29, 30
5322
XM_011511054.2
XP_011509356.1
29, 30
5322
XM_011511056.1
XP_011509358.1
19, 20
2942
XM_017003918.1
XP_016859407.1
34, 35
6132
XM_017003919.1
XP_016859408.1
34, 35
6126
XM_017003920.1
XP_016859409.1
36, 37
6343
XM_017003921.1
XP_016859410.1
31, 32
5666
XM_017003922.1
XP_016859411.1
31, 32
5562
XM_017003923.1
XP_016859412.1
29, 30
5316
XM_017003924.1
XP_016859413.1
28, 29
5214
XM_017003925.1
XP_016859414.1
30, 31
5570
XM_017003926.1
XP_016859415.1
30, 31
5428
XM_017003927.1
XP_016859416.1
30, 31
5468
XM_017003928.1
XP_016859417.1
30, 31
5378
XM_017003929.1
XP_016859418.1
30, 31
5460
XM_017003930.1
XP_016859419.1
30, 31
5268
XM_017003931.1
XP_016859420.1
30, 31
5366
XM_017003932.1
XP_016859421.1
27, 28
5019
XM_017003933.1
XP_016859422.1
29, 30
5245
XM_017003934.1
XP_016859423.1
29, 30
5140
XM_017003935.1
XP_016859424.1
28, 29
5078
XM_017003936.1
XP_016859425.1
28, 29
5032
XM_017003937.1
XP_016859426.1
23, 24
3739
GenBank mRNA
AB032255.1
-
25, 26
5065
AB040909.2
-
29, 30
5552
AK074895.1
-
8, 9
1810
AL080173.1
-
1, 2
132
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0