Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NXPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152745.2 | NP_689958.1 | 3 | 1309 | ||
GenBank mRNA | AB047362.1 | - | 3 | 1309 | ||
AB073665.1 | - | 3 | 1309 | |||
AK091655.1 | - | 3 | 767 | |||
AK095624.1 | - | 4 | 1308 | |||
AK296248.1 | - | 4 | 767 | |||
AK315141.1 | - | 3 | 877 | |||
BC047505.1 | - | 3 | 734 | 5313563 |