Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST6GALNAC6 SIRNA Assays ›
Gene Symbol:
ST6GALNAC6
Gene Name:
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
Gene Aliases:
SIAT7-F, SIAT7F, ST6GALNACVI
Chromosome Location:
Chr.9: 127885321 - 127906725 on Build GRCh38
UniGene ID:
Hs.109672
Species:
Human
siRNA ID
HSS121226
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST6GALNAC6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286999.1
NP_001273928.1
5
568
NM_001287000.1
NP_001273929.1
4
513
NM_001287001.1
NP_001273930.1
4
538
NM_001287002.1
NP_001273931.1
4
509
NM_013443.4
NP_038471.2
5
568
NR_104629.1
4
509
XM_011518610.2
XP_011516912.1
6
1309
XM_011518611.2
XP_011516913.1
5
1254
XM_017014658.1
XP_016870147.1
4
1158
XM_017014659.1
XP_016870148.1
6
1309
XM_017014660.1
XP_016870149.1
4
1158
XM_017014661.1
XP_016870150.1
5
1254
XM_017014662.1
XP_016870151.1
4
488
XM_017014663.1
XP_016870152.1
5
648
XM_017014664.1
XP_016870153.1
4
538
XM_017014665.1
XP_016870154.1
5
575
XM_017014666.1
XP_016870155.1
5
648
XM_017014667.1
XP_016870156.1
4
919
GenBank mRNA
AB035173.1
-
3
366
AJ507293.1
-
3
305
AK023900.1
-
4
464
AK057100.1
-
4
538
AK226174.1
-
4
394
AK310850.1
-
4
509
AY358672.1
-
4
474
BC006564.2
-
5
485
3009935
BC007802.2
-
5
512
4128833
BC016299.2
-
5
512
3507812
BG699392.1
-
5
583
CR457318.1
-
3
291
KU178617.1
-
4
393
KU178618.1
-
4
393
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
glycosylceramide metabolic process
glycosphingolipid metabolic process
glycoprotein metabolic process
oligosaccharide metabolic process
oligosaccharide biosynthetic process
cell-cell recognition
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0