Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 8, 9 | 1172 | ||
NM_001034838.2 | NP_001030010.1 | 7, 8 | 1019 | |||
NM_001278339.1 | NP_001265268.1 | 7, 8 | 1214 | |||
NM_001278340.1 | NP_001265269.1 | 8, 9 | 1301 | |||
NM_014592.3 | NP_055407.1 | 7, 8 | 1139 | |||
XM_017009407.1 | XP_016864896.1 | 8, 9 | 1143 | |||
GenBank mRNA | AF199597.1 | - | 7, 8 | 602 | ||
AK074390.1 | - | 7, 8 | 735 | |||
AK301775.1 | - | 7, 8 | 794 | |||
AY170821.1 | - | 7, 8 | 815 | |||
AY780424.1 | - | 8, 9 | 977 | |||
BC035032.1 | - | 7, 8 | 1144 | |||
BC050375.1 | - | 7, 8 | 906 | 5752207 | ||
DQ148476.1 | - | 7, 8 | 629 | |||
DQ148477.1 | - | 7, 8 | 602 | |||
DQ148478.1 | - | 8, 9 | 635 | |||
DQ148479.1 | - | 7, 8 | 677 |