Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 750 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1026 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 882 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1086 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1140 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 675 | |||
NM_021155.3 | NP_066978.1 | 7 | 1158 | |||
NR_026692.1 | 6 | 1258 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 775 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1100 | ||
AK293089.1 | - | 7 | 1114 | |||
AK304190.1 | - | 7 | 661 | |||
AK308595.1 | - | 4 | 701 | |||
AK313585.1 | - | 7 | 1159 | |||
AY042221.1 | - | 7 | 1091 | |||
AY042222.1 | - | 7 | 1073 | |||
AY042223.1 | - | 7 | 815 | |||
AY042224.1 | - | 6 | 383 | |||
AY042225.1 | - | 6 | 1019 | |||
AY042226.1 | - | 5 | 959 | |||
AY042227.1 | - | 6 | 683 | |||
AY042229.1 | - | 6 | 1191 | |||
AY042230.1 | - | 5 | 1119 | |||
AY042231.1 | - | 5 | 981 | |||
BC110615.2 | - | 5 | 1126 | 40032459 | ||
M98457.1 | - | 7 | 1132 |