Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015718.2 | NP_056533.1 | 10 | 1351 | ||
GenBank mRNA | AF190122.1 | - | 10 | 1351 | ||
AF229177.1 | - | 8 | 1090 |