Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251989.1 | NP_001238918.1 | 6, 7 | 670 | ||
NM_001251990.1 | NP_001238919.1 | 6, 7 | 594 | |||
NM_001251991.1 | NP_001238920.1 | 6, 7 | 578 | |||
NM_015895.4 | NP_056979.1 | 6, 7 | 802 | |||
XM_005249159.1 | XP_005249216.1 | 6, 7 | 841 | |||
XM_011514651.2 | XP_011512953.1 | 5, 6 | 2896 | |||
GenBank mRNA | AF067855.1 | - | 5, 6 | 488 | ||
AK021685.1 | - | 6, 7 | 670 | |||
AK222992.1 | - | 5, 6 | 461 | |||
AK315743.1 | - | 6, 7 | 783 | |||
AL558330.3 | - | 6, 7 | 785 | |||
BC005185.1 | - | 6, 7 | 725 | 4101771 | ||
BC005389.1 | - | 6, 7 | 777 | 3996265 | ||
BG777134.1 | - | 6, 7 | 552 | |||
BM699599.1 | - | 576 | ||||
CB131959.1 | - | 3, 4 | 292 |