Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RMDN1 SIRNA Assays ›
Gene Symbol:
RMDN1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
CGI-90, FAM82B, RMD-1, RMD1
Chromosome Location:
Chr.8: 86467116 - 86514348 on Build GRCh38
UniGene ID:
Hs.145386
Species:
Human
siRNA ID
HSS121698
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RMDN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286707.1
NP_001273636.1
8
872
NM_001286719.1
NP_001273648.1
8
911
NM_001317807.1
NP_001304736.1
9
780
NM_016033.2
NP_057117.2
9
1001
XM_005250944.3
XP_005251001.1
9
907
XM_005250947.3
XP_005251004.1
8
817
XM_005250948.3
XP_005251005.1
9
809
XM_006716573.3
XP_006716636.1
9
996
XM_011517090.2
XP_011515392.1
9
981
XM_011517091.2
XP_011515393.1
9
942
XM_011517092.2
XP_011515394.1
8
891
XM_011517093.2
XP_011515395.1
9
981
XM_011517094.2
XP_011515396.1
9
981
XM_011517095.2
XP_011515397.1
9
981
XM_011517096.2
XP_011515398.1
9
942
XM_011517097.2
XP_011515399.1
8
891
XM_011517098.2
XP_011515400.1
8
852
XM_017013518.1
XP_016869007.1
9
907
XM_017013519.1
XP_016869008.1
9
942
XM_017013520.1
XP_016869009.1
9
907
XM_017013521.1
XP_016869010.1
9
868
XM_017013524.1
XP_016869013.1
9
809
XM_017013525.1
XP_016869014.1
9
809
XM_017013526.1
XP_016869015.1
9
770
XM_017013527.1
XP_016869016.1
8
906
XM_017013528.1
XP_016869017.1
8
714
GenBank mRNA
AF151848.1
-
9
913
AK000672.1
-
9
909
AK297896.1
-
8
773
AK303122.1
-
8
763
AL561197.3
-
9
754
BC008509.1
-
7
565
BC009671.1
-
9
893
3896168
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0