Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040443.2 | NP_001035533.1 | 4 | 831 | ||
NM_001040444.2 | NP_001035534.1 | 4 | 507 | |||
NR_135322.1 | 3 | 709 | ||||
NR_135323.1 | 5 | 643 | ||||
GenBank mRNA | AB011031.1 | - | 4 | 480 | ||
AF155105.1 | - | 4 | 450 | |||
AK026228.1 | - | 4 | 471 | |||
AK225986.1 | - | 5 | 593 | |||
AK293242.1 | - | 4 | 831 | |||
AK300410.1 | - | 4 | 471 | |||
AL552215.3 | - | 4 | 639 | |||
BC017212.2 | - | 4 | 460 | 4054134 | ||
BX461254.2 | - | 3 | 339 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 14 | 2827 | ||
NR_135324.1 | 14 | 2866 |