Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016141.3 | NP_057225.2 | 4 | 489 | ||
GenBank mRNA | AF078849.1 | - | 4 | 465 | ||
AK074348.1 | - | 4 | 473 | |||
AK222573.1 | - | 4 | 424 | |||
AK222653.1 | - | 4 | 425 | |||
BC131620.1 | - | 4 | 435 | 40117391 | ||
BX649189.1 | - | 4 | 308 | |||
DA284201.1 | - | 4 | 489 |