Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 12 | 1134 | ||
NM_001135644.1 | NP_001129116.1 | 11 | 1281 | |||
NM_016221.3 | NP_057305.1 | 11 | 1113 | |||
XM_005268457.1 | XP_005268514.1 | 10 | 1007 | |||
XM_011537644.1 | XP_011535946.1 | 12 | 1017 | |||
XM_011537645.1 | XP_011535947.1 | 12 | 929 | |||
XM_011537646.1 | XP_011535948.1 | 12 | 1150 | |||
XM_011537647.1 | XP_011535949.1 | 12 | 973 | |||
GenBank mRNA | AF195120.1 | - | 11 | 1028 | ||
AK000299.1 | - | 11 | 1032 | |||
AK125973.1 | - | 11 | 1281 | |||
AK299517.1 | - | 11 | 1034 | |||
AK307757.1 | - | 12 | 1073 | |||
AL162085.1 | - | 1 | 63 | |||
KU178637.1 | - | 11 | 1011 | |||
KU178638.1 | - | 11 | 1011 | |||
KU178639.1 | - | 7 | 767 |