Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300888.1 | NP_001287817.1 | 5 | 643 | ||
NM_016500.3 | NP_057584.2 | 6 | 748 | |||
GenBank mRNA | BC001220.1 | - | 6 | 748 | 3528586 | |
BC051894.1 | - | 6 | 610 | 6528202 | ||
CD704296.1 | - | 5 | 537 |