Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 1 | 203 | ||
NM_001297749.1 | NP_001284678.1 | 1 | 203 | |||
NM_001297750.1 | NP_001284679.1 | 1 | 203 | |||
NM_001297751.1 | NP_001284680.1 | 1 | 203 | |||
NM_016511.3 | NP_057595.2 | 1 | 203 | |||
XM_011520687.2 | XP_011518989.1 | 1 | 203 | |||
GenBank mRNA | AF200949.1 | - | 1 | 141 | ||
AK075114.1 | - | 1 | 123 | |||
AK091989.1 | - | 1 | 154 | |||
AK291584.1 | - | 1 | 91 | |||
AK300979.1 | - | 1 | 90 | |||
AK313589.1 | - | 1 | 121 | |||
AY358587.1 | - | 1 | 203 | |||
BC039072.1 | - | 1 | 109 | 5178017 | ||
BC067746.1 | - | 1 | 123 | 30337675 | ||
BM920364.1 | - | 143 | ||||
BQ722745.1 | - | 1 | 132 | |||
BX339075.2 | - | 1 | 99 | |||
BX380212.2 | - | 1 | 60 |