Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002000.2 | NP_001002000.1 | 6 | 958 | ||
NM_001002001.2 | NP_001002001.1 | 6 | 899 | |||
NM_001002002.2 | NP_001002002.1 | 6 | 638 | |||
NM_001283021.1 | NP_001269950.1 | 7 | 1034 | |||
NM_001283022.1 | NP_001269951.1 | 5 | 879 | |||
NM_001283023.1 | NP_001269952.1 | 5 | 554 | |||
NM_016576.4 | NP_057660.2 | 5 | 879 | |||
NR_104265.1 | 6 | 982 | ||||
XM_005267740.3 | XP_005267797.1 | 6 | 764 | |||
XM_005267741.3 | XP_005267798.1 | 6 | 823 | |||
XM_005267742.3 | XP_005267799.1 | 6 | 642 | |||
XM_006720165.2 | XP_006720228.1 | 7 | 742 | |||
XM_017021356.1 | XP_016876845.1 | 7 | 958 | |||
XM_017021357.1 | XP_016876846.1 | 7 | 997 | |||
XM_017021358.1 | XP_016876847.1 | 7 | 1098 | |||
XM_017021359.1 | XP_016876848.1 | 7 | 741 | |||
XM_017021360.1 | XP_016876849.1 | 4 | 345 | |||
GenBank mRNA | AB032903.1 | - | 5 | 505 | ||
AF086919.1 | - | 6 | 674 | |||
AF135159.1 | - | 6 | 844 | |||
AF419346.1 | - | 6 | 748 | |||
AK226166.1 | - | 7 | 790 | |||
BC003053.1 | - | 5 | 828 | 3546620 | ||
BC008021.1 | - | 5 | 827 | 3537989 | ||
BC009832.2 | - | 6 | 556 | 3958714 | ||
BC093039.1 | - | 6 | 697 | 30347740 | ||
BX161436.1 | - | 6 | 879 | |||
BX161518.1 | - | 6 | 769 | |||
BX247993.1 | - | 5 | 477 | |||
CR457156.1 | - | 5 | 497 |