Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WT1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120546.1 | 2 | 1875 | |||
NR_120547.1 | 3 | 1533 | ||||
GenBank mRNA | BC002734.2 | - | 1 | 140 | ||
BC096708.1 | - | 1 | 490 | |||
BC098290.1 | - | 1 | 490 | |||
BC100673.1 | - | 1 | 490 | |||
DQ289488.1 | - | 2 | 2694 | |||
DQ289489.1 | - | 2 | 1874 | |||
DQ289490.1 | - | 2 | 1121 | |||
M60614.1 | - | 1 | 990 |