Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISYNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170938.1 | NP_001164409.1 | 10 | 1633 | ||
NM_001253389.1 | NP_001240318.1 | 8 | 1499 | |||
NM_016368.4 | NP_057452.1 | 11 | 1795 | |||
NR_045573.1 | 10 | 1601 | ||||
NR_045574.1 | 8 | 1465 | ||||
XM_011528059.1 | XP_011526361.1 | 10 | 1874 | |||
GenBank mRNA | AF086921.1 | - | 11 | 1613 | ||
AF207640.1 | - | 11 | 1607 | |||
AF220530.1 | - | 11 | 1627 | |||
AK001325.1 | - | 10 | 1581 | |||
AK021526.1 | - | 11 | 1627 | |||
AK092179.1 | - | 10 | 1499 | |||
AK092433.1 | - | 7 | 1714 | |||
AK093461.1 | - | 8 | 1414 | |||
AK296022.1 | - | 10 | 1463 | |||
AL137749.1 | - | 8 | 1499 | |||
BC004320.1 | - | 3 | 447 | |||
BC018952.2 | - | 11 | 1610 | 4301272 | ||
BC066902.1 | - | 11 | 1625 | 5262041 |