Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016470.7 | NP_057554.4 | 4 | 637 | ||
XM_011528851.1 | XP_011527153.1 | 5 | 1112 | |||
XM_011528853.2 | XP_011527155.1 | 4 | 681 | |||
XM_011528854.2 | XP_011527156.1 | 3 | 420 | |||
XM_017027873.1 | XP_016883362.1 | 5 | 1177 | |||
GenBank mRNA | AF151041.1 | - | 4 | 520 | ||
AF161517.1 | - | 4 | 520 | |||
AF217514.1 | - | 4 | 503 | |||
AK000476.1 | - | 4 | 541 | |||
AK315155.1 | - | 4 | 502 | |||
AL133000.1 | - | 4 | 522 | |||
BC064973.1 | - | 4 | 487 | 6151501 | ||
BI462161.1 | - | 4 | 646 |