Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184728.2 | NP_001171657.1 | 11 | 1434 | ||
NM_001184729.2 | NP_001171658.1 | 11 | 1401 | |||
NM_001184730.2 | NP_001171659.1 | 10 | 1296 | |||
NM_015937.5 | NP_057021.2 | 12 | 1602 | |||
NR_047691.1 | 12 | 1562 | ||||
NR_047692.1 | 11 | 1505 | ||||
NR_047693.1 | 11 | 1501 | ||||
NR_047694.1 | 11 | 1424 | ||||
NR_047695.1 | 9 | 1195 | ||||
XM_005260430.2 | XP_005260487.1 | 9 | 1073 | |||
XM_005260432.2 | XP_005260489.1 | 11 | 1396 | |||
GenBank mRNA | AB057724.1 | - | 12 | 1512 | ||
AF132940.1 | - | 12 | 1506 | |||
AK075469.1 | - | 12 | 1526 | |||
AK225517.1 | - | 11 | 1331 | |||
AK293294.1 | - | 11 | 1425 | |||
AK293686.1 | - | 11 | 1347 | |||
AK296063.1 | - | 11 | 1431 | |||
AK296139.1 | - | 11 | 1357 | |||
AK297830.1 | - | 7 | 1081 | |||
AK299013.1 | - | 9 | 1121 | |||
AK302093.1 | - | 10 | 1254 | |||
AK316358.1 | - | 12 | 1488 | |||
AL121742.1 | - | 12 | 1505 | |||
AY358588.1 | - | 12 | 1509 | |||
BC015022.1 | - | 12 | 1531 | 3921680 | ||
BC110892.1 | - | 12 | 1510 | |||
BC136827.1 | - | 12 | 1540 | 9020818 | ||
BC136828.1 | - | 12 | 1540 | 9020819 | ||
BX537612.1 | - | 9 | 2739 |