Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143780.2 | NP_001137252.1 | 8, 9 | 871 | ||
NM_001321240.1 | NP_001308169.1 | 7, 8 | 802 | |||
NM_016016.3 | NP_057100.1 | 8, 9 | 847 | |||
XM_017024737.1 | XP_016880226.1 | 7, 8 | 754 | |||
GenBank mRNA | AB209576.1 | - | 6, 7 | 1459 | ||
AF119864.1 | - | 7, 8 | 633 | |||
AF151827.1 | - | 8, 9 | 789 | |||
AF317711.1 | - | 7, 8 | 709 | |||
AK026060.1 | - | 8, 9 | 789 | |||
AK289357.1 | - | 8, 9 | 832 | |||
AK294085.1 | - | 7, 8 | 787 | |||
AL133584.1 | - | 8, 9 | 789 | |||
BC001398.2 | - | 8, 9 | 793 | 3051436 | ||
BC009330.2 | - | 8, 9 | 765 | 4126268 | ||
BC096819.1 | - | 8, 9 | 774 | 6109001 |