Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143780.2 | NP_001137252.1 | 10 | 1007 | ||
NM_001321240.1 | NP_001308169.1 | 9 | 938 | |||
NM_001321241.1 | NP_001308170.1 | 10 | 1001 | |||
NM_016016.3 | NP_057100.1 | 10 | 983 | |||
XM_011524880.2 | XP_011523182.1 | 10 | 982 | |||
XM_011524882.2 | XP_011523184.1 | 9 | 922 | |||
XM_011524883.2 | XP_011523185.1 | 9 | 924 | |||
XM_011524884.1 | XP_011523186.1 | 9 | 908 | |||
XM_017024737.1 | XP_016880226.1 | 9 | 890 | |||
GenBank mRNA | AB209576.1 | - | 8 | 1595 | ||
AF119864.1 | - | 9 | 769 | |||
AF151827.1 | - | 10 | 925 | |||
AF317711.1 | - | 9 | 845 | |||
AK026060.1 | - | 10 | 925 | |||
AK289357.1 | - | 10 | 968 | |||
AK294085.1 | - | 9 | 923 | |||
AL133584.1 | - | 10 | 925 | |||
BC001398.2 | - | 10 | 929 | 3051436 | ||
BC009330.2 | - | 10 | 901 | 4126268 | ||
BC096819.1 | - | 10 | 910 | 6109001 |