- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: C14orf166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016039.2 | NP_057123.1 | 6, 7 | 671 | ||
GenBank mRNA | AF087877.1 | - | 6, 7 | 702 | ||
AF100755.1 | - | 6, 7 | 702 | |||
AF151857.1 | - | 6, 7 | 702 | |||
AK312004.1 | - | 6, 7 | 635 | |||
AY817175.1 | - | 6, 7 | 541 | |||
BC001722.1 | - | 6, 7 | 623 | 3528725 | ||
BF594675.1 | - | 2 | 390 | |||
DQ525179.1 | - | 6, 7 | 706 |