Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERAP1 SIRNA Assays ›
Gene Symbol:
ERAP1
Gene Name:
endoplasmic reticulum aminopeptidase 1
Gene Aliases:
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
Chromosome Location:
Chr.5: 96759245 - 96935983 on Build GRCh38
UniGene ID:
Hs.666524
Species:
Human
siRNA ID
HSS122605
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040458.1
NP_001035548.1
8
1646
NM_001198541.1
NP_001185470.1
8
1404
NM_016442.3
NP_057526.3
8
1646
XM_005272015.4
XP_005272072.1
8
1597
XM_005272016.4
XP_005272073.1
8
1557
XM_011543480.2
XP_011541782.1
12
3866
XM_011543481.2
XP_011541783.1
12
3863
XM_011543484.2
XP_011541786.1
12
3862
XM_011543485.2
XP_011541787.1
11
3682
XM_011543486.2
XP_011541788.1
12
3866
XM_017009580.1
XP_016865069.1
13
3922
XM_017009581.1
XP_016865070.1
11
3708
XM_017009582.1
XP_016865071.1
8
1381
XM_017009583.1
XP_016865072.1
8
1539
GenBank mRNA
AB011097.2
-
7
1320
AF106037.1
-
8
1361
AF183569.1
-
8
1366
AF222340.1
-
8
1386
AK291636.1
-
8
1396
AK310535.1
-
7
821
AY028806.1
-
8
1396
AY028807.1
-
8
1396
AY358590.1
-
8
1384
BC030775.1
-
8
1398
4824919
JX962699.1
-
7
1299
JX962700.1
-
7
1299
JX962701.1
-
7
1299
JX962702.1
-
7
1299
KM357870.1
-
7
1299
KM357871.1
-
7
1299
KM357872.1
-
7
1299
KM357873.1
-
7
1299
KM357874.1
-
7
1299
KM357875.1
-
7
1299
KM357876.1
-
7
1299
KM357877.1
-
7
1299
KM357878.1
-
7
1299
KM357879.1
-
7
1299
KM357880.1
-
7
1299
KM357881.1
-
7
1299
KM357882.1
-
7
1299
KM357883.1
-
7
1299
KM357884.1
-
7
1299
KM357885.1
-
7
1299
KM357886.1
-
7
1299
KM357887.1
-
7
1299
KM357888.1
-
7
1299
KM357889.1
-
7
1299
KM357890.1
-
7
1299
KM357891.1
-
7
1299
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
membrane protein ectodomain proteolysis
regulation of blood pressure
response to bacterium
antigen processing and presentation of endogenous peptide antigen via MHC class I
peptide catabolic process
regulation of innate immune response
fat cell differentiation
positive regulation of angiogenesis
endopeptidase activity
aminopeptidase activity
interleukin-6 receptor binding
interleukin-1, Type II receptor binding
tumor necrosis factor receptor binding
protein binding
metalloexopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0