Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK12 SIRNA Assays ›
Gene Symbol:
CDK12
Gene Name:
cyclin dependent kinase 12
Gene Aliases:
CRK7, CRKR, CRKRS
Chromosome Location:
Chr.17: 39461486 - 39567560 on Build GRCh38
UniGene ID:
Hs.345028
Species:
Human
siRNA ID
HSS122612
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015083.2
NP_055898.1
13
3903
NM_016507.3
NP_057591.2
13
3903
XM_005257456.3
XP_005257513.1
13
3925
XM_005257458.4
XP_005257515.1
13
3928
XM_011524892.2
XP_011523194.1
12
3850
XM_011524893.2
XP_011523195.1
13
3928
XM_011524894.2
XP_011523196.1
13
3928
XM_011524895.2
XP_011523197.1
13
3928
XM_011524896.2
XP_011523198.1
13
3928
XM_011524897.2
XP_011523199.1
13
3928
XM_011524898.2
XP_011523200.1
13
3928
XM_011524899.2
XP_011523201.1
13
3928
XM_011524900.2
XP_011523202.1
13
3928
XM_011524901.2
XP_011523203.1
13
3928
XM_011524902.2
XP_011523204.1
13
3928
XM_011524903.2
XP_011523205.1
13
3928
XM_011524905.2
XP_011523207.1
13
3928
XM_011524906.2
XP_011523208.1
13
3928
XM_011524907.2
XP_011523209.1
13
3928
XM_017024744.1
XP_016880233.1
13
3928
XM_017024745.1
XP_016880234.1
13
3928
XM_017024746.1
XP_016880235.1
13
3928
XM_017024747.1
XP_016880236.1
13
3928
XM_017024748.1
XP_016880237.1
13
3928
XM_017024749.1
XP_016880238.1
13
3928
XM_017024750.1
XP_016880239.1
13
3928
XM_017024751.1
XP_016880240.1
13
3928
XM_017024752.1
XP_016880241.1
13
3928
XM_017024753.1
XP_016880242.1
13
3925
GenBank mRNA
AB020711.2
-
13
3653
AF227198.1
-
13
3375
AK302645.1
-
13
3900
BC140854.1
-
13
3362
9021722
BC150265.1
-
13
3659
9007217
BM557102.1
-
2
145
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
RNA splicing
regulation of MAP kinase activity
regulation of RNA splicing
protein autophosphorylation
regulation of cell cycle
phosphorylation of RNA polymerase II C-terminal domain
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
RNA polymerase II carboxy-terminal domain kinase activity
cyclin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0