Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017421.3 | NP_059117.3 | 7 | 968 | ||
XM_005267018.4 | XP_005267075.1 | 6 | 741 | |||
XM_006715500.3 | XP_006715563.1 | 8 | 1107 | |||
XM_011535898.2 | XP_011534200.1 | 7 | 865 | |||
XM_011535899.1 | XP_011534201.1 | 6 | 796 | |||
XM_017010933.1 | XP_016866422.1 | 8 | 965 | |||
GenBank mRNA | AF193016.1 | - | 7 | 905 | ||
AF351615.1 | - | 7 | 973 | |||
AK056955.1 | - | 7 | 966 | |||
AL136726.1 | - | 7 | 966 | |||
BC001534.1 | - | 7 | 939 | |||
BC015634.2 | - | 7 | 929 | |||
BC063463.1 | - | 7 | 944 | 5196692 |