Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018944.2 | NP_061817.1 | 3 | 527 | ||
XM_017028400.1 | XP_016883889.1 | 3 | 618 | |||
XM_017028401.1 | XP_016883890.1 | 3 | 618 | |||
XM_017028402.1 | XP_016883891.1 | 3 | 226 | |||
GenBank mRNA | AF231921.1 | - | 3 | 499 | ||
AF387845.1 | - | 3 | 497 | |||
AK075281.1 | - | 3 | 499 | |||
AK312073.1 | - | 3 | 618 | |||
BC042917.1 | - | 3 | 503 | 5934336 | ||
BU567844.1 | - | 1 | 94 | |||
BU616573.1 | - | 3 | 530 |