Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287813.2 | NP_001274742.1 | 20 | 1902 | ||
NM_001287814.2 | NP_001274743.1 | 17 | 1943 | |||
NM_001321773.1 | NP_001308702.1 | 18 | 2054 | |||
NM_018967.4 | NP_061840.1 | 19 | 2129 | |||
NR_135794.1 | 19 | 2109 | ||||
NR_135795.1 | 19 | 2124 | ||||
NR_135796.1 | 19 | 2155 | ||||
NR_135797.1 | 21 | 2299 | ||||
XM_011517548.2 | XP_011515850.1 | 18 | 2239 | |||
XM_017013579.1 | XP_016869068.1 | 21 | 5058 | |||
XM_017013580.1 | XP_016869069.1 | 19 | 1652 | |||
GenBank mRNA | AJ003030.1 | - | 17 | 1579 | ||
AK314084.1 | - | 20 | 1839 | |||
AL161971.1 | - | 19 | 2024 | |||
BC075072.2 | - | 17 | 1494 | 30915592 | ||
BC104829.1 | - | 18 | 1576 | 8143832 | ||
BC143695.1 | - | 18 | 1602 |