Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018982.4 | NP_061855.1 | 5, 6 | 657 | ||
NR_036639.1 | 5, 6 | 657 | ||||
NR_036640.1 | 4, 5 | 437 | ||||
NR_135075.1 | 3, 4 | 374 | ||||
XM_017001499.1 | XP_016856988.1 | 4, 5 | 415 | |||
XM_017001500.1 | XP_016856989.1 | 4, 5 | 478 | |||
XM_017001501.1 | XP_016856990.1 | 3, 4 | 259 | |||
GenBank mRNA | AK000823.1 | - | 3, 4 | 322 | ||
AK300240.1 | - | 4, 5 | 519 | |||
AK315183.1 | - | 5, 6 | 635 | |||
AL713668.1 | - | 4, 5 | 368 | |||
BC064590.1 | - | 5, 6 | 623 | 5749036 | ||
BI757212.1 | - | 5, 6 | 654 | |||
BX405963.2 | - | 5, 6 | 649 | |||
CN308348.1 | - | 5, 6 | 658 |