Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242581.1 | NP_001229510.1 | 8 | 1338 | ||
NM_018993.3 | NP_061866.1 | 7 | 1078 | |||
XM_005260731.2 | XP_005260788.1 | 9 | 1534 | |||
XM_005260733.2 | XP_005260790.1 | 5 | 886 | |||
XM_006723574.3 | XP_006723637.1 | 10 | 1516 | |||
XM_006723575.3 | XP_006723638.1 | 9 | 1302 | |||
XM_006723577.2 | XP_006723640.1 | 8 | 1151 | |||
XM_011529255.2 | XP_011527557.2 | 8 | 1611 | |||
XM_011529257.1 | XP_011527559.1 | 8 | 1207 | |||
XM_011529258.2 | XP_011527560.1 | 8 | 1219 | |||
XM_011529259.2 | XP_011527561.1 | 7 | 1131 | |||
XM_017027887.1 | XP_016883376.1 | 11 | 1857 | |||
XM_017027888.1 | XP_016883377.1 | 12 | 1929 | |||
XM_017027889.1 | XP_016883378.1 | 7 | 1458 | |||
XM_017027890.1 | XP_016883379.1 | 9 | 1289 | |||
XM_017027891.1 | XP_016883380.1 | 9 | 1327 | |||
XM_017027892.1 | XP_016883381.1 | 7 | 1142 | |||
XM_017027893.1 | XP_016883382.1 | 8 | 1636 | |||
GenBank mRNA | AB060339.1 | - | 7 | 1042 | ||
AK094884.1 | - | 8 | 1339 | |||
AL136924.1 | - | 6 | 959 |