Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318163.1 | NP_001305092.1 | 8 | 964 | ||
NM_001318164.1 | NP_001305093.1 | 8 | 1153 | |||
NM_019042.4 | NP_061915.2 | 8 | 1177 | |||
XM_005250462.3 | XP_005250519.1 | 9 | 1203 | |||
XM_011516336.2 | XP_011514638.1 | 9 | 1198 | |||
XM_017012367.1 | XP_016867856.1 | 9 | 1131 | |||
XM_017012368.1 | XP_016867857.1 | 7 | 1044 | |||
GenBank mRNA | AB067484.1 | - | 8 | 891 | ||
AK091283.1 | - | 8 | 1157 | |||
AK128629.1 | - | 8 | 1139 | |||
BC011396.1 | - | 8 | 1133 | 3870711 |