Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 18, 19 | 1579 | ||
XM_011511359.1 | XP_011509661.1 | 18, 19 | 1576 | |||
XM_011511361.1 | XP_011509663.1 | 17, 18 | 1333 | |||
XM_011511362.1 | XP_011509664.1 | 17, 18 | 1447 | |||
GenBank mRNA | AB209993.1 | - | 18, 19 | 1498 | ||
AK001858.1 | - | 6, 7 | 373 | |||
AK025611.1 | - | 17, 18 | 1347 | |||
AK057926.1 | - | 18, 19 | 1440 | |||
AK290722.1 | - | 18, 19 | 1486 | |||
AK309040.1 | - | 18, 19 | 1486 | |||
BC028609.1 | - | 18, 19 | 1552 | 4826022 | ||
BC093018.1 | - | 18, 19 | 1469 | 30337330 |