Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 23 | 2471 | ||
NR_125340.1 | 20 | 2218 | ||||
XM_005244931.2 | XP_005244988.1 | 22 | 2411 | |||
XM_006711192.2 | XP_006711255.1 | 24 | 2381 | |||
XM_006711193.3 | XP_006711256.1 | 24 | 2513 | |||
XM_006711194.3 | XP_006711257.1 | 11 | 12509 | |||
XM_011509246.2 | XP_011507548.1 | 23 | 2332 | |||
XM_011509247.1 | XP_011507549.1 | 22 | 2272 | |||
XM_011509248.2 | XP_011507550.1 | 23 | 2265 | |||
XM_011509249.2 | XP_011507551.1 | 24 | 2570 | |||
XM_017000470.1 | XP_016855959.1 | 24 | 2459 | |||
XM_017000471.1 | XP_016855960.1 | 23 | 2321 | |||
XM_017000472.1 | XP_016855961.1 | 19 | 1945 | |||
GenBank mRNA | AK000061.1 | - | 9 | 1008 | ||
AK024832.1 | - | 2 | 355 | |||
AK091207.1 | - | 20 | 2032 | |||
BC074735.2 | - | 6 | 857 |