Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127458.1 | NP_001120930.1 | 5 | 548 | ||
NM_001323561.1 | NP_001310490.1 | 5 | 556 | |||
NM_001323562.1 | NP_001310491.1 | 5 | 563 | |||
NM_001323563.1 | NP_001310492.1 | 5 | 931 | |||
NM_001323564.1 | NP_001310493.1 | 5 | 557 | |||
NM_019095.5 | NP_061968.1 | 5 | 869 | |||
NR_136617.1 | 4 | 783 | ||||
XM_005260738.4 | XP_005260795.1 | 5 | 533 | |||
GenBank mRNA | AF241784.1 | - | 5 | 788 | ||
AL832419.1 | - | 5 | 616 | |||
BC069010.1 | - | 4 | 706 | 5092704 | ||
BM563835.1 | - | 5 | 551 | |||
BP368984.1 | - | 5 | 548 | |||
BU852955.1 | - | 6 | 576 | |||
DQ386730.1 | - | 5 | 695 |