Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243746.1 | NP_001230675.1 | 12 | 1929 | ||
NM_017565.3 | NP_060035.2 | 11 | 1703 | |||
NR_027751.1 | 11 | 1130 | ||||
XM_006721959.2 | XP_006722022.1 | 11 | 1075 | |||
GenBank mRNA | AB545606.1 | - | 11 | 1703 | ||
AF447872.1 | - | 5 | 1516 | |||
AK056789.1 | - | 12 | 1543 | |||
AK298071.1 | - | 12 | 1571 | |||
AL133105.1 | - | 9 | 812 | |||
AY358197.1 | - | 12 | 1703 | |||
BC036222.1 | - | 11 | 1094 | |||
BC136686.1 | - | 11 | 1592 | 9020676 | ||
BC136689.1 | - | 12 | 1592 | 9020679 | ||
CB530015.1 | - | 1 | 586 |