Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017589.3 | NP_060059.1 | 2 | 344 | ||
XM_011542876.2 | XP_011541178.1 | 2 | 378 | |||
XM_011542878.2 | XP_011541180.1 | 2 | 378 | |||
XM_011542879.2 | XP_011541181.1 | 2 | 378 | |||
XM_011542880.2 | XP_011541182.1 | 2 | 378 | |||
GenBank mRNA | AJ271351.1 | - | 1 | 149 | ||
BC031045.1 | - | 2 | 330 | 5271706 | ||
BX099301.1 | - | 2 | 344 |