Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304725.1 | NP_001291654.1 | 3 | 418 | ||
NM_017655.5 | NP_060125.4 | 3 | 715 | |||
GenBank mRNA | AB073737.1 | - | 3 | 531 | ||
AK000082.1 | - | 2 | 274 | |||
AK225941.1 | - | 3 | 554 | |||
AK307422.1 | - | 3 | 418 | |||
BC036075.1 | - | 3 | 715 | 5296057 |