Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135589.2 | NP_001129061.1 | 13 | 1700 | ||
NM_017686.3 | NP_060156.1 | 13 | 1700 | |||
XM_005270959.2 | XP_005271016.1 | 13 | 1700 | |||
XM_011541621.2 | XP_011539923.1 | 13 | 1700 | |||
XM_011541622.1 | XP_011539924.1 | 13 | 1588 | |||
XM_011541623.2 | XP_011539925.1 | 13 | 1700 | |||
XM_011541624.2 | XP_011539926.1 | 14 | 2143 | |||
XM_017001531.1 | XP_016857020.1 | 14 | 1722 | |||
XM_017001532.1 | XP_016857021.1 | 14 | 2143 | |||
GenBank mRNA | AK000149.1 | - | 13 | 1603 | ||
BC013132.1 | - | 13 | 1583 | 3448678 | ||
BX459360.2 | - | 2 | 560 |