Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104554.1 | NP_001098024.1 | 9 | 1473 | ||
NM_017705.3 | NP_060175.3 | 9 | 1183 | |||
XM_005254495.4 | XP_005254552.1 | 7 | 969 | |||
XM_011521720.2 | XP_011520022.1 | 10 | 1344 | |||
XM_011521722.2 | XP_011520024.1 | 10 | 1385 | |||
XM_011521723.2 | XP_011520025.1 | 8 | 1062 | |||
XM_011521725.2 | XP_011520027.1 | 7 | 881 | |||
XM_011521727.2 | XP_011520029.1 | 8 | 1096 | |||
XM_017022360.1 | XP_016877849.1 | 12 | 1584 | |||
XM_017022361.1 | XP_016877850.1 | 9 | 1201 | |||
XM_017022362.1 | XP_016877851.1 | 11 | 1504 | |||
XM_017022363.1 | XP_016877852.1 | 8 | 1080 | |||
XM_017022364.1 | XP_016877853.1 | 9 | 1280 | |||
XM_017022366.1 | XP_016877855.1 | 8 | 1032 | |||
XM_017022368.1 | XP_016877857.1 | 6 | 739 | |||
GenBank mRNA | AK000197.1 | - | 9 | 1473 | ||
AY424283.1 | - | 9 | 1473 | |||
BC039234.1 | - | 10 | 1183 | 4608045 |
Gene Symbol: LOC145694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132969.1 | 2 | 1521 | |||
GenBank mRNA | DB570961.1 | - | 1 | 81 |
Set Membership: |
![]() |