Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017762.2 | NP_060232.2 | 13, 14 | 1470 | ||
XM_005254508.2 | XP_005254565.1 | 12, 13 | 1349 | |||
XM_011521737.2 | XP_011520039.1 | 13, 14 | 1440 | |||
XM_011521738.2 | XP_011520040.1 | 13, 14 | 1440 | |||
XM_017022384.1 | XP_016877873.1 | 13, 14 | 1440 | |||
GenBank mRNA | AK000320.1 | - | 13, 14 | 1470 | ||
AK296926.1 | - | 8, 9 | 883 | |||
AL832137.1 | - | 1, 2 | 2036 | |||
AL832603.1 | - | 13, 14 | 1465 | |||
BC063296.1 | - | 12, 13 | 1378 | |||
BC094750.1 | - | 14, 15 | 1610 | |||
BC121171.1 | - | 7, 8 | 680 | |||
BC121172.1 | - | 7, 8 | 680 | |||
KJ534883.1 | - | 13, 14 | 1372 | |||
KJ535030.1 | - | 13, 14 | 1378 | |||
KJ535071.1 | - | 12, 13 | 1287 | |||
KJ535075.1 | - | 13, 14 | 1371 | |||
KJ535083.1 | - | 12, 13 | 1247 |