Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXK SIRNA Assays ›
Gene Symbol:
PXK
Gene Name:
PX domain containing serine/threonine kinase like
Gene Aliases:
MONaKA
Chromosome Location:
Chr.3: 58332868 - 58426127 on Build GRCh38
UniGene ID:
Hs.190544
Species:
Human
siRNA ID
HSS123488
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289098.1
NP_001276027.1
1, 2
196
NM_017771.4
NP_060241.2
1, 2
196
XM_005265250.4
XP_005265307.1
1, 2
196
XM_005265252.4
XP_005265309.1
1, 2
196
XM_005265255.3
XP_005265312.1
1, 2
196
XM_005265256.3
XP_005265313.1
1, 2
196
XM_017006675.1
XP_016862164.1
1, 2
196
XM_017006676.1
XP_016862165.1
1, 2
196
XM_017006677.1
XP_016862166.1
1, 2
196
XM_017006678.1
XP_016862167.1
1, 2
196
XM_017006679.1
XP_016862168.1
1, 2
196
XM_017006680.1
XP_016862169.1
1, 2
196
XM_017006686.1
XP_016862175.1
1, 2
154
XM_017006687.1
XP_016862176.1
1, 2
152
XM_017006692.1
XP_016862181.1
1, 2
191
XM_017006693.1
XP_016862182.1
1, 2
191
XM_017006694.1
XP_016862183.1
1, 2
85
XM_017006695.1
XP_016862184.1
1, 2
191
GenBank mRNA
AB209688.1
-
1, 2
175
AF399753.1
-
1, 2
97
AK000342.1
-
1, 2
194
AY274811.1
-
1, 2
196
AY847222.1
-
1, 2
99
BC008943.2
-
1, 2
177
BC014479.1
-
1, 2
112
4856139
DA296189.1
-
1, 2
197
DQ124707.1
-
1, 2
97
DQ124708.1
-
1, 2
97
KF774202.1
-
1, 2
206
KF774203.1
-
1, 2
206
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inflammatory response
negative regulation of ATPase activity
regulation of membrane potential
negative regulation of ion transport
modulation of synaptic transmission
nucleotide binding
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0