Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 8 | 1059 | ||
NM_001166270.1 | NP_001159742.1 | 7 | 916 | |||
NM_017815.2 | NP_060285.2 | 8 | 1051 | |||
GenBank mRNA | AK000431.1 | - | 8 | 1020 | ||
AK026016.1 | - | 8 | 1005 | |||
AK057779.1 | - | 6 | 846 | |||
AY927599.1 | - | 3 | 299 | |||
BC001916.1 | - | 8 | 1048 | 2905474 | ||
BC002554.2 | - | 8 | 982 | 3139198 | ||
BX247981.1 | - | 5 | 857 | |||
BX247995.1 | - | 7 | 868 | |||
BX248256.1 | - | 7 | 883 |