Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017820.4 | NP_060290.3 | 14 | 1642 | ||
XM_011518810.1 | XP_011517112.1 | 16 | 2014 | |||
XM_011518814.1 | XP_011517116.1 | 8 | 964 | |||
XM_011518815.1 | XP_011517117.1 | 5 | 648 | |||
XM_017014861.1 | XP_016870350.1 | 17 | 2158 | |||
XM_017014862.1 | XP_016870351.1 | 15 | 1899 | |||
XM_017014863.1 | XP_016870352.1 | 15 | 1857 | |||
XM_017014864.1 | XP_016870353.1 | 14 | 2219 | |||
XM_017014865.1 | XP_016870354.1 | 14 | 1866 | |||
XM_017014866.1 | XP_016870355.1 | 13 | 1522 | |||
XM_017014867.1 | XP_016870356.1 | 13 | 1563 | |||
GenBank mRNA | AB061798.1 | - | 8 | 1078 | ||
AK055004.1 | - | 4 | 774 | |||
AK091390.1 | - | 8 | 1793 | |||
AK094438.1 | - | 14 | 1542 |